Learn More
Miele Professional™ Mobile unit
Product Code. 15430555 Shop All Miele Products
Click to view available options
:
capacity: 15 DIN mesh trays; model: levels
Product Code. 15430555
Brand: Miele Professional™ 7765760
Mobile unit E 900-5/2 for the optimum cleaning of instruments on 5 levels
  • Loading capacity: 15 DIN mesh trays
  • The loading areas for the individual levels are different
  • Magnetic strip for mobile unit recognition (incl. magnets ML)
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Product Type Mobile Unit for Mesh Trays
Color Stainless Steel
Holds 15 DIN mesh trays10 SPRI mesh trays
For Use With (Equipment) Washer-disinfectors, laboratory
Depth (Metric) 790 mm
Height (Metric) 605 mm
Width (Metric) 640 mm