Learn More
Miele Professional™ Mobile unit
Shop All Miele Products
Click to view available options
:
Dimensions (D x W X H): 790 x 619 x 421mm
Description
- The mobile unit can also be used without the upper level
- Magnetic strip for automatic mobile unit recognition
- Lower level equipped with a total of 115 nozzles with clips
Specifications
Specifications
| Material | Stainless Steel |
| Color | Stainless Steel |
| For Use With (Equipment) | Large-capacity washer-disinfectors, laboratory |
| Depth (Metric) | 790 mm |
| Height (Metric) | 421 mm |
| Width (Metric) | 619 mm |