Learn More
Memmert™ MobileALERT for IF75
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF75 Incubator
Unit Size:
Each
Description
- MobileALERT
- Notification by SMS in case of any error or alarm of the device
- Requires option H6
Specifications
Specifications
| For Use With (Equipment) | IF75 Incubator |
| Type | Alerting Device |