Learn More
Memmert™ MobileALERT for IN750
Product Code. 17597895 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN750 Incubator
Unit Size:
Each
Product Code. 17597895
Brand: Memmert™ IN750C3
Only null left
Add to basket
Add to basket
MobileALERT provides a notification by SMS with potential free contact error option.
  • MobileALERT
  • Notification by SMS in case of any error or alarm of the device
  • Requires option H6
Specifications
For Use With (Equipment) IN750 Incubator
Type Alerting Device