missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOBKL1A Monoclonal Antibody (OTI2H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502280S
This item is not returnable.
View return policy
Description
MOBKL1A Monoclonal antibody specifically detects MOBKL1A in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| MOBKL1A | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16864 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9296 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| OTI2H4 | |
| Unconjugated | |
| Atp6v1f | |
| 1110003E08Rik; AU015450; B230364F10; MATS2; MGC33910; MOB kinase activator 1B; mob1 homolog 1A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; mob1A; MOB1B; MOB4A; Mobkl1a; mps one binder kinase activator-like 1A | |
| Full length human recombinant protein of human ATP6V1F produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction