missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOBKL2B Antibody (OTI6C6), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72748R
This item is not returnable.
View return policy
Description
MOBKL2B Monoclonal specifically detects MOBKL2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| MOBKL2B | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MOB3B | |
| Full length recombinant protein of human MOBKL2B (NP_079037) produced in HEK293T cells. | |
| 0.1 mL | |
| 79817.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI6C6 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Em:AL163192.1, FLJ13204, FLJ23916, Mob1 homolog 2b, MOB1, Mps One Binder kinase activator-like 2B (yeast), MOB3BMGC32960, monopolar spindle 1 binding, MOB1, domain containing, mps one binder kinase activator-like 2B, Protein Mob3B | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction