missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557179
This item is not returnable.
View return policy
Description
Immunogen sequence: SQKPAKEGPR LSKNQKYSEH FSIHCCPPFT FLNSKKEIVD RKYSICKSGC FYQKKEEDWI CCACQKTR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
MOBP may play a role in compacting or stabilizing the myelin sheath, possibly by binding the negatively charged acidic phospholipids of the cytoplasmic membrane.
Specifications
| MOBP | |
| Polyclonal | |
| Unconjugated | |
| Mobp | |
| AI835053; AI839848; Mobp; MOBP155; MOBP170; MOBP69; MOBP73; Mobp81; Mobp81p; myelin associated oligodendrocyte basic protein; myelin-associated oligodendrocyte basic protein; myelin-associated oligodendrocytic basic protein; Myelin-associated/Oligodendrocytic Basic Protein-81; R74645; RNMOBP81P; Unknown (protein for MGC:128384) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4336 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13875 | |
| Mobp | |
| Recombinant protein corresponding to Human MOBP. Recombinant protein control fragment (Product #RP-96435). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction