missing translation for 'onlineSavingsMsg'
Learn More

Moesin Antibody (MSN/492), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-47916AF488

Product Code. 18733323

  • € 588.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Moesin Monoclonal antibody specifically detects Moesin in Human, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF, Gene Knock-Out
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Moesin
Monoclonal
Alexa Fluor 488
Membrane-organizing extension spike protein, moesin
Recombinant full-length human Moesin protein (Uniprot: P26038 )
0.1 mL
Cell Biology, Cellular Markers, Stem Cell Markers
4478
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
MSN/492
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Rat (Negative)
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.