missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moesin Antibody (rMSN/492), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08923JF549
This item is not returnable.
View return policy
Description
Moesin Monoclonal antibody specifically detects Moesin in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Moesin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Membrane-organizing extension spike protein, moesin | |
| Recombinant full-length human Moesin protein (Uniprot: P26038) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| 4478 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| rMSN/492 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction