missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moesin Antibody (SPM562), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34798CL1
This item is not returnable.
View return policy
Description
Moesin Monoclonal antibody specifically detects Moesin in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),CyTOF,Gene Knock-Out,Immunocytochemistry/Immunofluorescence
Specifications
| Moesin | |
| Monoclonal | |
| CoraFluor 1 | |
| Membrane-organizing extension spike protein, moesin | |
| Moesin Purified from uterus (Uniprot: P26038) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| 4478 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), CyTOF | |
| SPM562 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction