missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Moesin Polyclonal Antibody

Description
Immunogen sequence: KKAQQELEEQ TRRALELEQE RKRAQSEAEK LAKERQEAEE AKEALLQASR DQKKTQEQLA LEMAELTARI SQLEMARQKK ESEAVEWQQK AQMVQEDLEK T Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 98%.
Moesin, a member of the talin-4. 1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.
Specifications
Specifications
| Antigen | Moesin |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MSN |
| Gene Accession No. | O35763, P26038, P26041 |
| Gene Alias | C78546; epididymis luminal protein 70; HEL70; Membrane-organizing extension spike protein; MGC137877 protein; moesin; MSN |
| Gene Symbols | MSN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?