missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOG Antibody (013), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89453JF549
This item is not returnable.
View return policy
Description
MOG Monoclonal antibody specifically detects MOG in Human samples. It is validated for ELISA
Specifications
| MOG | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 013 | |
| ELISA | |
| MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human MOG (NP_996532.2; Gly 30-Tyr 149). | |
| 0.1 mL | |
| Cellular Markers | |
| 4340 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction