Learn More
MOG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12690-1-AP-20UL
 View more versions of this product
Product Code. 16890544
  • € 145.00 / 20µL
Description
The product of this gene is a membrane protein expressed on the oligodendrocyte cell surface and the outermost surface of myelin sheaths. Due to this localization, it is a primary target antigen involved in immune-mediated demyelination. This protein may be involved in completion and maintece of the myelin sheath and in cell-cell communication.
Specifications
Specifications
MOG
Polyclonal
Unconjugated
MOG
MOG, MOGIG 2, myelin oligodendrocyte glycoprotein
Rabbit
Antigen Affinity Chromatography
RUO
17441, 24558, 4340
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16653, Q61885, Q63345
Mog
MOG Fusion Protein Ag3273
20 μL
Primary
Mouse, Rat, Human