Learn More
MOG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12690-1-AP-20UL
This item is not returnable.
View return policy
Description
The product of this gene is a membrane protein expressed on the oligodendrocyte cell surface and the outermost surface of myelin sheaths. Due to this localization, it is a primary target antigen involved in immune-mediated demyelination. This protein may be involved in completion and maintece of the myelin sheath and in cell-cell communication.Specifications
| MOG | |
| Polyclonal | |
| Unconjugated | |
| MOG | |
| MOG, MOGIG 2, myelin oligodendrocyte glycoprotein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17441, 24558, 4340 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16653, Q61885, Q63345 | |
| Mog | |
| MOG Fusion Protein Ag3273 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |