Learn More
MOGAT3 Antibody (3F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00346606-M05
Product Code. 18575274
  • € 589.00 / 0.10mg
Description
MOGAT3 Monoclonal antibody specifically detects MOGAT3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MOGAT3
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
acyl coenzyme A:monoacylglycerol acyltransferase 3, Acyl-CoA:monoacylglycerol acyltransferase 3, DC72-acylglycerol O-acyltransferase 3, DGAT2L7MGC119203, Diacylglycerol acyltransferase 2-like protein 7, Diacylglycerol O-acyltransferase candidate 7, EC 2.3.1.20, EC 2.3.1.22, MGAT3, MGC119204, monoacylglycerol O-acyltransferase 3hDC7
MOGAT3 (NP_835470, 59 a.a. ∽ 107 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YLVWLYVDWDTPNQGGRRSEWIRNRAIWRQLRDYYPVKLVKTAELPPDR
0.1 mg
Primary
Human
Purified
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
3F7
Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
NP_835470
Mouse
IgG purified
RUO
346606
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG1 κ