Learn More
Invitrogen™ 5(6)-CFDA (5-(and-6)-Carboxyfluorescein Diacetate), mixed isomers
Product Code. 11534237
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11534237 100 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11534237 Supplier Invitrogen™ Supplier No. C195
Edge
A cell-permeant esterase substrate that can serve as a viability probe
  • Measures both enzymatic activity, which is require to activate its fluorescence, and cell-membrane integrity, which is required for intracellular retention of their fluorescent product
  • Upon hydrolysis by intracellular esterases, this AM ester also yields carboxyfluorescein
  • As compared with fluorescein, carboxyfluorescein contains extra negative charges and is therefore better retained in cells
Cell Adhesion, Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Proliferation, Cell Tracing and Tracking, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Cellular Toxicology Assays, Chemotaxis and Cell Migration, Drug Discovery and Development, General Cell Tracing, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Ionic Homeostasis and Signaling, Target-Based ADME/Tox Assays, pH Detection
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer -5°C to -30°C.
Product Type 5(6)-CFDA
Dye Type Other Label(s) or Dye(s)
Quantity 100 mg
Reagent Type CFDA & Related Compounds
Shipping Condition Room Temperature