Learn More
Invitrogen™ FocalCheck™ Fluorescent Microspheres Kit, 6 μm, mounted on slides
Product Code. 10286612
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 10286612
Brand: Invitrogen™ F24633
Beads are pre-mounted on slides for confocal microscope calibration
Our popular FocalCheck microspheres have been available in suspension for several years. Now, in response to customer requests, we offer FocalCheck beads pre-mounted on microscope slides. The 6 μm FocalCheck Fluorescent Microsphere Kit features mounted samples of our three best-selling color configurations: fluorescent green/orange/dark red ring stains, fluorescent green ring stain/blue throughout and fluorescent dark red ring stain/green throughout.
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Calibration Type Confocal Microscope Calibration
Content And Storage Store at room temperature and protect from light.
Color Orange, Dark Red, Blue, Green
Product Type Fluorescent Microspheres Kit
Format Mounted on Slide
Product Line FocalCheck
Quantity 1 Kit
Shipping Condition Room Temperature
Diameter (Metric) 6 μm