missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GST Tag Polyclonal Antibody, Alexa Fluor™ 488, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen A11131
Additional Details : Weight : 0.00450kg
Description
GST Tag Polyclonal antibody specifically detects GST Tag in Tag samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Order Info
Shipping Condition: Wet Ice
Specifications
GST Tag | |
Polyclonal | |
Alexa Fluor 488 | |
activator protein 1; AP1; AP-1; cancer/testis antigen 48; c-Jun; CT48; deafness, X-linked 7; DFN7; enhancer-binding protein AP1; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; glutathione S-transferase mu 1; Glutathione S-transferase P; glutathione S-transferase pi 1; Glutathione-S-Transferase; gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GSTP1-1; GTH4; GTM1; H-B; HB subunit 4; HEL-S-22; JUN; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; MU; MU-1; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; PI; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; SLC21A19 | |
GST | |
500 μL | |
Primary | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Rabbit | |
IgG fraction | |
RUO | |
4° C, store in dark | |
Liquid | |
Tag |
For Research Use Only. Not for use in diagnostic procedures.