Learn More
Invitrogen™ Escherichia coli BioParticles™ Opsonizing Reagent
Product Code. 10483072
Click to view available options
Quantity:
1 unit
Unit Size:
Each
Product Code. 10483072
Brand: Invitrogen™ E2870
The Escherichia coli BioParticles™ opsonizing agent is derived from highly purified rabbit polyclonal IgG antibodies.
The Escherichia coli BioParticles™ opsonizing agent is derived from highly purified rabbit polyclonal IgG antibodies and specifically designed to enhance the uptake of the Escherichia coli BioParticals™ conjugates.
See User Manual for solubility instructions.
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5°C to -30°C).
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Opsonizing Reagent
Dye Type Unconjugated
Form Solid
Product Line BioParticles
Quantity 1 unit
Shipping Condition Room Temperature
Species E. coli