Learn More
Invitrogen™ LysoSensor™ Yellow/Blue dextran, 10,000 MW, Anionic, Fixable
Description
LysoSensor Yellow/Blue dextran can be used to measure the pH of acidic organelles, such as lysosomes. This dextran conjugate is taken up by the cells through endocytosis. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue dextran is ∽3.9.
Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Ionic Homeostasis & Signaling, Lysosomes, Organelle Tracing, pH Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Yellow, Blue |
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Dextran |
| Emission | 335/452 |
| Product Line | LysoSensor |
| Quantity | 5 mg |
| Shipping Condition | Room Temperature |
| Sub Cellular Localization | Lysosomes |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.