Learn More
Invitrogen™ Qtracker™ 585 Cell Labeling Kit
Product Code. 10551675
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10551675 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10551675 Supplier Invitrogen™ Supplier No. Q25011MP
Includes
100μL Qtracker nanocrystals, Component A (2μM in 50mm borate buffer, pH 8.3) and 100μL Qtracker carrier, Component B (phosphate buffered saline, pH 7.2).
Designed for loading cells grown in culture with highly fluorescent Qdot nanocrystals
  • Use proprietary targeting peptide to deliver green-fluorescent Qdot 525 nanocrystals into cytoplasm of live cells
  • Once inside cells, Qtracker labels provide intense, stable fluorescence that can be traced through several generations, and are not transferred to adjacent cells in population
  • Qtracker 525 label has Ex/Em (405-485/525)nm
  • Provide intense, stable fluorescence that can be traced through several generations
  • Excellent tools for long-term tracking or imaging studies of live cells, including migration, motility, morphology, and other cell function assays
  • Cytoplasmic delivery by this mechanism is not mediated by specific enzyme; therefore, no cell-type specificity has been observed
  • Delivery is typically accomplished in less than 1 hour
  • Compatible with serum-sensitive cells; intense fluorescence is maintained in complex cellular environments and under various biological conditions including changes in intracellular pH, temperature, and metabolic activity
  • Observe labeled cells using extensive continuous illumination, without photobleaching and degradation problems often associated with organic dyes
  • Distributed in vesicles in cytoplasm, and are inherited by daughter cells for at least six generations
  • Can be seen up to a week after delivery in some cell lines
  • Do not leak out of intact cells to be taken up by adjacent cells in population
  • Qtracker reagent-labeled live cells can be easily monitored on variety of platforms, including flow cytometry, fluorescence/confocal microscopy, fluorescence microplate readers, and high-content imaging systems
  • Cytotoxicity of materials has been tested in variety of cell lines including CHO, HeLa, U-118, 3T3, HUVEC, and Jurkat cells: Labeling appears to exert minimal impact on cellular surface marker expression, cell proliferation, cellular enzyme activity, and cell motility
  • Post-labeling, researchers have demonstrated wide variety of applications including cell co-culture and cell assembly into heterotypic assemblies, multilineage differentiation, trans-differentiation versus cell fusion, embryonic and mesenchymal stem cell tracking, and cell migration dynamics
Cell Analysis, Cell Structure, Cell Tracing and Tracking, Cellular Imaging, Endosomes, General Cell Tracing, Immunofluorescence (IF), Lysosomes, Neuronal Tracing, Organelle Tracing
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Contains 100 μl Qtracker™ nanocrystals, Component A (2 μM in 50 mM borate buffer, pH 8.3) and 100 μl Qtracker™ carrier,Component B (phosphate buffered saline, pH 7.2). Store at 2–6°C. Do not freeze. Stable for at least 6 months.
Quantity 1 kit
Product Line QTRACKER, Qdot
Reagent Type Cell Labeling Reagents
Shipping Condition Room Temperature
Product Type Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.