missing translation for 'onlineSavingsMsg'
Learn More

Molidustat, MedChemExpress

Product Code. 16447997
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447997

Brand: Medchem Express HY12654

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Molidustat (BAY 85-3934) is a novel inhibitor of hypoxia-inducible factor prolyl hydroxylase (HIF-PH) with mean IC50 values of 480 nM for PHD1, 280 nM for PHD2, and 450 nM for PHD3.

Molidustat (BAY 85-3934) is a novel inhibitor of hypoxia-inducible factor prolyl hydroxylase (HIF-PH) with mean IC50 values of 480 nM for PHD1, 280 nM for PHD2, and 450 nM for PHD3.

Specifications

Chemical Name or Material Molidustat
Physical Form Solid
CAS 1154028-82-6
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H14N8O2
For Use With (Application) COVID-19-immunoregulation
Synonym BAY 85-3934
Solubility Information DMSO : 5 mg/mL (15.91 mM; Need ultrasonic)
SMILES O=C1C(N2N=NC=C2)=CNN1C3=NC=NC(N4CCOCC4)=C3
Molecular Weight (g/mol) 314.3
Formula Weight 314.3
Percent Purity 99.26%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.