Learn More
Molindone hydrochloride, MedChemExpress
Product Code. 16458648
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16458648
Brand: Medchem Express HYB1017
MedChemExpress Molindone hydrochloride (EN-1733A) is a therapeutic antipsychotic, used in the treatment of schizophrenia, works by blocking the effects of dopamine in the brain, leading to diminished psychoses.
Molindone hydrochloride (EN-1733A) is a therapeutic antipsychotic, used in the treatment of schizophrenia, works by blocking the effects of dopamine in the brain, leading to diminished psychoses.
Specifications
Chemical Name or Material Molindone hydrochloride
Physical Form Solid
CAS 15622-65-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H25ClN2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym EN-1733A
Solubility Information H2O : 100 mg/mL (319.66 mM; Need ultrasonic) ∣DMSO : 17.5 mg/mL (55.94 mM; Need ultrasonic and warming)
SMILES O=C1C2=C(NC(C)=C2CC)CCC1CN3CCOCC3.[H]Cl
Molecular Weight (g/mol) 312.83
Formula Weight 312.83
Percent Purity 99.5%
Grade Research
Show More Show Less
Research purposes only