Learn More
Molybdate Standard: MoO4(-2) @ 1000 μg/mL in H2O, VHG™
Product Code. 30003006
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 30003006
Brand: VHG Labs VHGIMOLB100
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Molybdate Standard: MoO4(-2) @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Molybdate
CAS 11116-47-5
Methods Ion Chromatography
Analyte or Component Names Molybdate
Shipping Condition Room Temperature
Molecular Formula (Mo O4)2-
Certifications/Compliance ISO 17034
Synonym MoO42−
SMILES [O-][Mo](=O)(=O)[O-]
IUPAC Name Molybdate
Solvent or Matrix Water
Molecular Weight (g/mol) 159.94
Quantity 100 mL
Formula Weight 159.94 g/mol