missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum Standard: Mo @ 1000 μg/mL in NH4OH, VHG™
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
Description
Molybdenum Standard: Mo @ 1000 μg/mL in NH4OH
Specifications
Specifications
| Chemical Name or Material | Molybdenum |
| CAS | 7439-98-7 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Molybdenum |
| Molecular Formula | Mo |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Molybdenum, 3N5, Amperit 105.054, Amperit 106.2, MChVL, Metco 63, Molybdenum element, NSC 600660, NSC 600661, NSC 600665, NSC 603570, NSC 603571, NSC 603572, SGC 15, TMO 10, TMO 30, TMO 50, TMOIO, TsM1, Molybdaen, Molybdän |
| InChI Formula | InChI=1S/Mo |
| SMILES | [Mo] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction