missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum Standard: Mo @ 10000 μg/mL in 5% HNO3, tr. HF

Product Code. 30321745
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321745 250 mL 250mL
30317236 100 mL 100mL
30325654 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30321745 Supplier VHG Labs Supplier No. VHGTMONF250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molybdenum
CAS 7439-98-7
Analyte or Component Names Molybdenum
Shipping Condition Room Temperature
Molecular Formula Mo
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Molybdenum, 3N5, Amperit 105.054, Amperit 106.2, MChVL, Metco 63, Molybdenum element, NSC 600660, NSC 600661, NSC 600665, NSC 603570, NSC 603571, NSC 603572, SGC 15, TMO 10, TMO 30, TMO 50, TMOIO, TsM1, Molybdaen
InChI Formula InChI=1 S/Mo
Solution Type Single Solution
SMILES [Mo]
Element Mo
IUPAC Name Molybdenum
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 95.95
Quantity 250 mL
Formula Weight 97.9054
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.