missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MON1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35801-100ul
This item is not returnable.
View return policy
Description
MON1A Polyclonal antibody specifically detects MON1A in Human samples. It is validated for ELISA,Western Blot
Specifications
| MON1A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FLJ97088, MON1 homolog A (yeast), SAND1MGC13272, vacuolar fusion protein MON1 homolog A | |
| Recombinant protein of human MON1A. | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84315 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction