missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Monastrol
GREENER_CHOICE

Product Code. 18074852
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18074852

Brand: Tocris Bioscience™ 1305/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of mitotic kinesin Eg5

Potent, cell-permeable, small molecule mitosis inhibitor that does not interact with tubulin. Arrests cells in mitosis and specifically inhibits the motility of the mitotic kinesin Eg5, a motor protein required for mitotic spindle formation and maintenance (IC50 = 14 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 254753-54-3
Molecular Formula C14H16N2O3S

Specifications

Quantity 50 mg
Formula Weight 292.35
Purity >98%
Molecular Weight (g/mol) 292.353
Chemical Name or Material Monastrol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.