missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey C3a ELISA Kit
Click to view available options
:
96 Tests
Description
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
Specifications
Specifications
| Assay Range | 0.16 to 10 ng/mL |
| Assay Sensitivity | 0.09 ng/mL |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Immunoassay Kit Format | ELISA Microplate (Dismountable) |
| Product Type | ELISA |
| Sample Type | Plasma, Serum |
| Target Species | Monkey |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Interassay CV | <10 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction