missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-2 Monkey ELISA Kit
For the quantitative detection of monkey IL-2.
€ 634.00 - € 3950.00
Specifications
Accession Number | Q29615,Q865Y1 |
---|---|
Assay Range | 18.8-1,200 U/mL |
Assay Sensitivity | 9.1 U/mL |
Product Type | ELISA Kit |
Sample Type | Amniotic Fluid, Plasma, Serum, Supernatant, Urine, Whole Blood |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-2
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15522087
|
Invitrogen™
BMS643-2 |
96 Tests |
€ 634.00
96 tests |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15552087
|
Invitrogen™
BMS643-2TWO |
2 x 96 Tests |
€ 854.00
Pack of 2 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15532087
|
Invitrogen™
BMS643-2FIVE |
5 x 96 Tests |
€ 2010.00
Pack of 5 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15542087
|
Invitrogen™
BMS643-2TEN |
10 x 96 Tests |
€ 3950.00
Pack of 10 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
The Monkey Interleukin 2 (Mk IL-2) ELISA quantitates Mk IL-2 in monkey serum or cell culture medium. The assay will exclusively recognize both natural and recombinant monkey IL-2. Principle of the method The Monkey IL-2 solid-phase sandwich E
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistencySpecifications
Q29615,Q865Y1 | |
9.1 U/mL | |
Amniotic Fluid, Plasma, Serum, Supernatant, Urine, Whole Blood | |
ELISA | |
5% | |
HRP | |
Amniotic Fluid, 50 μL; Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL; Urine,50 μL; Whole Blood,50 μL | |
Aldesleukin, T cell growth factor, TCGF | |
3 hr. 30 min. |
18.8-1,200 U/mL | |
ELISA Kit | |
Monkey | |
Colorimetric Microplate Reader | |
7% | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |