missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus A29 Antibody (0036), Biotin , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18814B
This item is not returnable.
View return policy
Description
Monkeypox Virus A29 Monoclonal antibody specifically detects Monkeypox Virus A29 in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Monkeypox Virus A29 | |
| Monoclonal | |
| Biotin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 0036 | |
| PBS | |
| This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A29 Protein expressed from HEK293 cells. | |
| 0.1 mL | |
| Primary | |
| Store at 4&drg;C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction