missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus A29L Antibody (0027), Janelia Fluor™ 646 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18270JF646
This item is not returnable.
View return policy
Description
Monkeypox Virus A29L Monoclonal antibody specifically detects Monkeypox Virus A29L in Virus samples. It is validated for Western Blot, ELISA, ELISA (Matched Antibody Pair)
Specifications
| Monkeypox Virus A29L | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 0027 | |
| 50mM Sodium Borate | |
| Recombinant Monkeypox Virus A29L Protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction