missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus A29L Antibody (0032), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18272
This item is not returnable.
View return policy
Description
Monkeypox Virus A29L Monoclonal antibody specifically detects Monkeypox Virus A29L in Virus samples. It is validated for Western Blot, ELISA (Matched Antibody Pair)
Specifications
| Monkeypox Virus A29L | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Recombinant Monkeypox Virus A29L Protein | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 32 | |
| Western Blot 1:500-1:5000, ELISA, ELISA Capture (Matched Antibody Pair) 1:250-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction