missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus A29L Humanized Recombinant Human Monoclonal Antibody (P1A2)
Monkeypox Virus A29L Humanized Recombinant Human Monoclonal Antibody (P1A2)
Brand: Invitrogen MA554748
This item is not returnable.
View return policy
Description
Monkeypox Virus A29L Humanized Recombinant Monoclonal Antibody for ELISA
Specifications
| Monkeypox Virus A29L Humanized | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| VHH-hFc |
| ELISA | |
| P1A2 | |
| Unconjugated | |
| A29L | |
| Recombinant MPXV A29L. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction