missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monkeypox Virus A29L Humanized Recombinant Human Monoclonal Antibody (P2B8)

Product Code. 30132116
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132116 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30132116 Supplier Invitrogen™ Supplier No. MA554757

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

Monkeypox Virus A29L is a gene that is found in the genome of the Monkeypox virus (MPXV). MPXV is a member of the Orthopoxvirus genus, which also includes the variola virus (which causes smallpox) and the vaccinia virus (used in smallpox vaccination). The A29L gene encodes a protein that is involved in various aspects of the viral life cycle, including viral replication, assembly, and immune evasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monkeypox Virus A29L Humanized
Applications ELISA
Classification Recombinant Monoclonal
Clone P2B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias A29L
Host Species Human
Immunogen Recombinant MPXV A29L.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype VHH-hFc
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.