missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus A35 Antibody (0017), DyLight 594 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18811DL594
This item is not returnable.
View return policy
Description
Monkeypox Virus A35 Monoclonal antibody specifically detects Monkeypox Virus A35 in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Monkeypox Virus A35 | |
| Monoclonal | |
| DyLight 594 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 0017 | |
| 50mM Sodium Borate | |
| This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A35 and expressed from HEK293 cells. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction