missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus B21R Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26104JF635
This item is not returnable.
View return policy
Description
Monkeypox Virus B21R Polyclonal antibody specifically detects Monkeypox Virus B21R in Virus samples. It is validated for Western Blot
Specifications
| Monkeypox Virus B21R | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Monkeypox Virus B21R | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 635 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction