missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus B6R Antibody (004) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 150.00 - € 462.00
Specifications
| Antigen | Monkeypox Virus B6R |
|---|---|
| Clone | 004 |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634345
|
Novus Biologicals
NBP3-23525-20ul |
20 μL |
€ 150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614064
|
Novus Biologicals
NBP3-23525-100ul |
100 μL |
€ 462.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Monkeypox Virus B6R Monoclonal antibody specifically detects Monkeypox Virus B6R in Virus samples. It is validated for Western Blot, ELISASpecifications
| Monkeypox Virus B6R | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| IgG | |
| Protein A purified |
| 004 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Monkeypox Virus B6R (Uniprot#: QNP13760.1; Tyr18-His279). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title