Learn More
Monkeypox Virus B6R Antibody (007) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-23527-100ul
 View more versions of this product
Product Code. 18641494
  • € 462.00 / 100µL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
Monkeypox Virus B6R Monoclonal antibody specifically detects Monkeypox Virus B6R in Virus samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monkeypox Virus B6R
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
This antibody was obtained from a rabbit immunized with purified, recombinant Monkeypox Virus B6R (Uniprot#: QNP13760.1; Tyr18-His279).
100 μL
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA, ELISA
007
Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Sandwich ELISA Capture : 1:250-1:2000
Rabbit
Protein A purified
RUO
Virus