missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus B6R Antibody (007) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23527-20ul
This item is not returnable.
View return policy
Description
Monkeypox Virus B6R Monoclonal antibody specifically detects Monkeypox Virus B6R in Virus samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| Monkeypox Virus B6R | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Monkeypox Virus B6R (Uniprot#: QNP13760.1; Tyr18-His279). | |
| 20 μL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, ELISA | |
| 007 | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Sandwich ELISA Capture : 1:250-1:2000 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction