missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus E8L Antibody (14) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-30751JF525
This item is not returnable.
View return policy
Description
Monkeypox Virus E8L Monoclonal antibody specifically detects Monkeypox Virus E8L in Virus samples. It is validated for ELISA,Western Blot,Lateral Flow
Specifications
| Monkeypox Virus E8L | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot, Lateral Flow | |
| 14 | |
| 50mM Sodium Borate | |
| Produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Monkeypox Virus E8L (RefSeq#: QNP13700.1; Met1-Thr275) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction