Learn More
Novus Biologicals™ Monkeypox Virus E8L Antibody (16) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30753AF488
Product Code. 30042664
  • € 507.00 / 0.10mL
Description
Monkeypox Virus E8L Monoclonal antibody specifically detects Monkeypox Virus E8L in Virus samples. It is validated for ELISA,Western Blot,Sandwich ELISA Capture
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monkeypox Virus E8L
Monoclonal
Alexa Fluor 488
Mouse
Protein A purified
RUO
Virus
Purified
ELISA, Western Blot, Sandwich ELISA Capture
16
50 mM Sodium Borate
Produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Monkeypox Virus E8L (RefSeq#: QNP13700.1; Met1-Thr275)
0.1 mL
Primary
Store at 4°C in the dark.