missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus Envelope H3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-40577AF594
This item is not returnable.
View return policy
Description
Monkeypox Virus Envelope H3 Polyclonal antibody specifically detects Monkeypox Virus Envelope H3 in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Monkeypox Virus Envelope H3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Produced in rabbits immunized with purified, recombinant Monkeypox Virus Envelope H3 (Uniprot#: QNP13688.1; Met1-Pro278). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| Alexa Fluor 594 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction