missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human PLAP ELISA Kit (Colorimetric)

Product Code. 30239194 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30239194

Brand: Novus Biologicals™ NBP339346

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.312 - 20 ng/mL (example only; lot dependent)
Assay Sensitivity 0.114 ng/mL (example only; lot dependent)
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Other Biological Fluids
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 9373
Gene Symbol PLAA
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym DOA1, FLJ12699, phospholipase A-2-activating protein, phospholipase A2-activating protein, PLAP
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.