missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mono-Methyl-Histone H1.1 (Lys21) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121430
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
| Mono-Methyl-Histone H1.1 (Lys21) | |
| Polyclonal | |
| Unconjugated | |
| H1-1 | |
| H1 histone family, member 1; H1.1; H1-1; H1A; H1F1; HIST1; HIST1H1A; histone 1, H1a; histone cluster 1, H1a; histone H1.1; Histone H1a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3024 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q02539 | |
| H1-1 | |
| Synthesized peptide derived from Human Histone H1.1 protein (18-30aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction