missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monoacylglycerol Lipase Recombinant Rabbit Monoclonal Antibody (HL1938)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547228
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Monoglyceride lipase functions together with hormone-sensitive lipase to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides.
Specifications
| Monoacylglycerol Lipase | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O35678, Q99685 | |
| MGLL | |
| Recombinant fragment of human Monoglyceride lipase. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1938 | |
| Unconjugated | |
| MGLL | |
| AA589436; HUK5; HU-K5; lysophospholipase homolog; Lysophospholipase-like; MAGL; Mgl; Mgl2; MGLL; Monoacylglycerol lipase; monoglyceride lipase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11343, 23945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction