missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monobenzylethylendiamine, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Monobenzylethylendiamine
Specifications
Specifications
| Chemical Name or Material | Monobenzylethylendiamine |
| CAS | 9-4-4152 |
| Analyte or Component Names | Monobenzylethylendiamine |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C9 H14 N2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzylpenicillin Benzathine Imp. A (EP), Monobenzylethylendiamine, Benzylpenicillin Benzathine Impurity A |
| InChI Formula | InChI=1S/C9H14N2/c10-6-7-11-8-9-4-2-1-3-5-9/h1-5,11H,6-8,10H2 |
| SMILES | NCCNCc1ccccc1 |
| IUPAC Name | N'-benzylethane-1,2-diamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction