Learn More
Monocrotaline, MedChemExpress
Product Code. p-200098151
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494877 50 mg 50mg
16449136 100 mg 100mg
16421516 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16494877 Supplier Medchem Express Supplier No. HYN0750
MedChemExpress Monocrotaline is an pyrrolizidine alkaloid extracted from the seeds of the Crotalaria spectabilis plant to induce pulmonary hypertension in rodents.
Monocrotaline is an pyrrolizidine alkaloid extracted from the seeds of the Crotalaria spectabilis plant to induce pulmonary hypertension in rodents.
Specifications
Chemical Name or Material Monocrotaline
CAS 315-22-0
Color White
Purity Grade Notes Research
Molecular Formula C16H23NO6
Quantity 50 mg
Synonym Crotaline
Solubility Information DMSO : 27.78 mg/mL (85.38 mM; Need ultrasonic) ∣H2O : 2 mg/mL (6.15 mM; Need ultrasonic)
SMILES O=C(O[C@]1([H])CCN2[C@]1([H])C(CO3)=CC2)[C@H](C)[C@@](C)(O)[C@@](C)(O)C3=O
Molecular Weight (g/mol) 325.36
Formula Weight 325.36
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only