missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monocyte/Granulocyte Antibody (74-22-15), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28241IR
This item is not returnable.
View return policy
Description
Monocyte/Granulocyte Monoclonal specifically detects Monocyte/Granulocyte in Mouse, Porcine samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Monocyte/Granulocyte | |
| Monoclonal | |
| DyLight 755 | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Porcine SWC3a | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 74-22-15 | |
| Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| IL8 | |
| Fresh dd miniature swine thymocytes | |
| 0.25 mL | |
| Angiogenesis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3576 | |
| Mouse, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction