missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoglyceride Lipase Antibody (OTI2B11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72759H
This item is not returnable.
View return policy
Description
Monoglyceride Lipase Monoclonal specifically detects Monoglyceride Lipase in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Monoglyceride Lipase | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| MGLL | |
| Full length recombinant protein of human Monoglyceride Lipase (NP_009214) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI2B11 | |
| Western Blot, Immunohistochemistry | |
| HU-K5EC 3.1.1.23, Lysophospholipase homolog, Lysophospholipase-like, MAGL, MGLHUK5, Monoacylglycerol lipase, monoglyceride lipase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 11343.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction