missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Suberate, TRC

Product Code. 30464889
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30464889

Brand: Toronto Research Chemicals TRCM54259025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monomethyl Suberate
CAS 3946-32-5
Molecular Formula C9H16O4
Quantity 25 g
Synonym Octanedioic Acid, Monomethyl Ester, Suberic Acid Methyl Ester, Suberic Acid, Monomethyl Ester, 8-Methoxy-8-oxooctanoic Acid, Methyl 1,8-Octanedionate, Methyl 7-Carboxyheptanoate
InChI Formula InChI=1S/C9H16O4/c1-13-9(12)7-5-3-2-4-6-8(10)11/h2-7H2,1H3,(H,10,11)
SMILES COC(=O)CCCCCCC(=O)O
IUPAC Name 8-methoxy-8-oxooctanoic acid
Molecular Weight (g/mol) 188.22
Formula Weight 188.1049
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.